Learn More
Proteintech Group Inc CoraLite488-conjugated PLOD3 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48860058100UL
Encompass_Preferred
The PLOD3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLOD3. This antibody recognizes human antigen. The PLOD3 antibody has been validated for the following applications: IF analysis. This antibody is coralite488.
Catalog No. 50-225-6892