Learn More
Proteintech Group Inc CoraLite488-conjugated TMEM173/STING Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48819851100UL
Encompass_Preferred
The TMEM173/STING antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMEM173/STING. This antibody recognizes human antigen. The TMEM173/STING antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite488.
Catalog No. 50-225-6854