Learn More
Proteintech Group Inc CoraLite488 TUNEL Assay Apoptosis Detection Kit(green)
SDP
Catalog No. 502258713 Shop All Proteintech Group Inc Products
Encompass_Preferred
A distinctive feature of apoptosis is degradation of DNA. The DNA fragments tend to be vary in length in integer multiples of 180 bp-200 bp, which is shown as a ladder-like pattern in agarose gel electrophoresis. This kit uses the TUNEL method, using Terminal Deoxynucleotidyl Transferase (TdT) to catalyze the incorporation of CL488-dUTP at the 3'-OH end of the broken DNA of apoptotic cells. CL488-dUTP-labeled DNA can be directly observed with a fluorescence microscope or quantified with a flow cytometer. The TUNEL method can selectively detect apoptotic cells, but not necrotic cells or cells with DNA strand breaks caused by irradiation and drug treatment.
This item is not returnable. View return policy
Catalog No. 50-225-8713 Supplier Proteintech Group Inc Supplier No. PF0000650T
A distinctive feature of apoptosis is degradation of DNA. The DNA fragments tend to be vary in length in integer multiples of 180 bp-200 bp, which is shown as a ladder-like pattern in agarose gel electrophoresis. This kit uses the TUNEL method, using Terminal Deoxynucleotidyl Transferase (TdT) to catalyze the incorporation of CL488-dUTP at the 3'-OH end of the broken DNA of apoptotic cells. CL488-dUTP-labeled DNA can be directly observed with a fluorescence microscope or quantified with a flow cytometer. The TUNEL method can selectively detect apoptotic cells, but not necrotic cells or cells with DNA strand breaks caused by irradiation and drug treatment.