Learn More
Proteintech Group Inc CoraLite594-conjugated c-SRC Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59460315100UL
Encompass_Preferred
The c-SRC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human c-SRC. This antibody recognizes human, hamster, mouse antigen. The c-SRC antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7623