Learn More
Proteintech Group Inc CoraLite594-conjugated CD13 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466211100UL
Encompass_Preferred
The CD13 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD13. This antibody recognizes human antigen. The CD13 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite594.
Catalog No. 50-225-7727