Learn More
Proteintech Group Inc CoraLite594-conjugated CD206 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59460143100UL
Encompass_Preferred
The CD206 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human CD206. This antibody recognizes human antigen. The CD206 antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7574