Learn More
Proteintech Group Inc CoraLite594-conjugated EIF4E Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466655100UL
Encompass_Preferred
The EIF4E antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF4E. This antibody recognizes human, mouse, rat antigen. The EIF4E antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite594.
Catalog No. 50-225-7824