Learn More
Proteintech Group Inc CoraLite594-conjugated Fibronectin Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466042100UL
Encompass_Preferred
The Fibronectin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Fibronectin. This antibody recognizes human, mouse, rat antigen. The Fibronectin antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite594.
Catalog No. 50-225-7668