Learn More
Proteintech Group Inc CoraLite594-conjugated HSP90 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59460318100UL
Encompass_Preferred
The HSP90 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HSP90. This antibody recognizes human, mouse, rat, pig antigen. The HSP90 antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7626