Learn More
Proteintech Group Inc CoraLite594-conjugated NSE Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466150100UL
Encompass_Preferred
The NSE antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human NSE. This antibody recognizes human, mouse, rat, pig antigen. The NSE antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7710