Learn More
Proteintech Group Inc CoraLite594-conjugated NUDT21 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466335100UL
Encompass_Preferred
The NUDT21 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NUDT21. This antibody recognizes human, mouse antigen. The NUDT21 antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7756