Learn More
Proteintech Group Inc CoraLite594-conjugated P62,SQSTM1 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466184100UL
Encompass_Preferred
The P62,SQSTM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P62,SQSTM1. This antibody recognizes human antigen. The P62,SQSTM1 antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7716