Learn More
Proteintech Group Inc CoraLite594-conjugated RAB5A Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466339100UL
Encompass_Preferred
The RAB5A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB5A. This antibody recognizes human, pig antigen. The RAB5A antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7757