Learn More
Proteintech Group Inc CoraLite594-conjugated RPS3 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466046100UL
Encompass_Preferred
The RPS3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human RPS3. This antibody recognizes human, mouse, rat antigen. The RPS3 antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7672