Learn More
Proteintech Group Inc CoraLite594-conjugated S100A4 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466489100UL
Encompass_Preferred
The S100A4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A4. This antibody recognizes human, mouse antigen. The S100A4 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite594.
Catalog No. 50-225-7796