Learn More
Proteintech Group Inc CoraLite594-conjugated SOD2 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466474100UL
Encompass_Preferred
The SOD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOD2. This antibody recognizes human, mouse, rat, pig antigen. The SOD2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite594.
Catalog No. 50-225-7789