Learn More
Proteintech Group Inc CoraLite594-conjugated TBR1 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL59466564100UL
Encompass_Preferred
The TBR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBR1. This antibody recognizes human, rat, mouse, pig antigen. The TBR1 antibody has been validated for the following applications: IF analysis. This antibody is coralite594.
Catalog No. 50-225-7812