Learn More
Proteintech Group Inc CoraLite647-conjugated PD-1/CD279 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL64766220100UL
Encompass_Preferred
The PD-1/CD279 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PD-1/CD279. This antibody recognizes human, rat, mouse antigen. The PD-1/CD279 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite647.
Catalog No. 50-225-8123