Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coralyne chloride | 38989-38-7 | MFCD00011938 | 98.3% | 399.87 g/mol | C22H22ClNO4 | 5 MG
SDP

Catalog No. 5000229912
Encompass_Preferred

Coralyne chloride is a protoberberine alkaloid used in research as a topoisomerase I poison and as a precursor for DNA-binding fluorescent probes. It is supplied as a high-purity, light yellow to yellow solid intended for laboratory applications.

  • Acts as a topoisomerase I poison.
  • Suitable for synthesizing coralyne derivatives and DNA-binding fluorescent probes.
  • Supplied as a light yellow to yellow solid.
  • High purity (98.3%) for research use.
  • Molecular formula C22H22ClNO4; molecular weight 399.87 g/mol.
  • Store solid at 4°C; in solution follow recommended storage conditions.

Catalog No. 50-002-29912 Supplier Medchemexpress LLC Supplier No. HY1185815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coralyne chloride is a protoberberine alkaloid used in research as a topoisomerase I poison and as a precursor for DNA-binding fluorescent probes. It is supplied as a high-purity, light yellow to yellow solid intended for laboratory applications.

  • Acts as a topoisomerase I poison.
  • Suitable for synthesizing coralyne derivatives and DNA-binding fluorescent probes.
  • Supplied as a light yellow to yellow solid.
  • High purity (98.3%) for research use.
  • Molecular formula C22H22ClNO4; molecular weight 399.87 g/mol.
  • Store solid at 4°C; in solution follow recommended storage conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.