Learn More
CPC Scientific Pyr-Thr-Phe-Gln-Tyr-Ser-Arg-Gly-Trp-Thr-Asn-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific INSE003B
Encompass
Corazonin is a cardioaccelerating peptide, which has been isolated from the corpus cardiacum of the American cockroach, Periplaneta americana. It is the most potent insect cardioactive neuropeptide.ONE-LETTER SEQUENCE: Pyr-TFQYSRGWTN-NH2MOLECULAR FORMULA: C62H84N18O18MOLECULAR WEIGHT:1369.46STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [122929-08-2], RESEARCH AREA: NeuropeptidesREFERENCES: J.A.Veenstra, FEBS Lett., 250, 231 (1989)
Catalog No. 50-210-9606