Learn More
StemExpress Cord Blood CD34+ Stem/Progenitor Cells, Single Donor, Frozen, 0.5 million/vial
SDP
Catalog No. 502126518
Encompass
Mononuclear cells are separated from umbilical cord blood by means of a density gradient centrifugation protocol. Human cord blood CD34 depleted mononuclear cells are isolated from the mononuclear cell fraction by removing the CD34+ population by immunomagnetic separation. The CD34 depleted mononuclear cells contain less than 0.1% CD34+ cells. The presence of CD34+ cells is assessed by flow cytometry prior to cryopreservation. The isolated MNCs can be used to isolate subpopulations or used as a control for donor matched CD34+ stem cell experiments (Please request the CD34 depleted flow-through fraction upon ordering CD34+ stem cells).
Catalog No. 50-212-6518 Supplier StemExpress Supplier No. CB3400.5C
Mononuclear cells are separated from umbilical cord blood by means of a density gradient centrifugation protocol. Human cord blood CD34 depleted mononuclear cells are isolated from the mononuclear cell fraction by removing the CD34+ population by immunomagnetic separation. The CD34 depleted mononuclear cells contain less than 0.1% CD34+ cells. The presence of CD34+ cells is assessed by flow cytometry prior to cryopreservation. The isolated MNCs can be used to isolate subpopulations or used as a control for donor matched CD34+ stem cell experiments (Please request the CD34 depleted flow-through fraction upon ordering CD34+ stem cells).