Learn More
Enzo Life Sciences Cord Factor (endotoxin-free grade) (1mg). CAS: 61512-20-7
SDP
Supplier:  Enzo Life Sciences ALX581210M001
Encompass_Preferred
Immunomodulatory compound / adjuvant. Induces activation of macrophages independent of TLR2 or TLR4. Alternative name: Trehalose 6,6'-dimycolate, TDM, 6,6'-Di-O-mycoloyl-alpha,alpha-trehalose. Formulation: Lyophilized. Purity: Single spot (TLC). Solubility: Soluble in chloroform:methanol:water (90:10:1) (5mg/ml), hexane or isopropanol. Long Term Storage: +4°C.
Catalog No. 50-200-9584