Learn More
ACCU-SCOPE™ Cord Hangers
SDP
Catalog No. 01000940 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Type:
Cord Hangers
Catalog No. Type
01-000-940 Cord Hangers
1 options
Catalog No. 01-000-940 Supplier ACCU-SCOPE™ Supplier No. 3251
Add to Cart
Add to Cart
Cord hanger compatible with 3000 series, 3000-LED series, 3012 series, 3012-LED series, EXC-350 series microscopes.
  • Plastic cord hanger with mounting screws
  • Allows easy cord storage when microscope is not in use.
  • Compatible with ACCU-SCOPE 3000 series, 3000-LED series, 3012 series, 3012-LED series, EXC-350 series microscopes
Specifications
Type Cord Hangers
Autoclavable Not Autoclavable
Color Black
Sterility Non-sterile