Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Cord Set, 13A, 125V, 16/3, USA

Catalog No. NC9465982 Shop All Thermo Scientific Products
Encompass_Preferred

Cord Set, 13A, 125V, 16/3, USA.

Catalog No. NC9465982 Supplier Thermo Scientific Supplier No. 430326
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cord Set, 13A, 125V, 16/3, USA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.