Learn More
Beckman Coulter Umbilical/Wall-Plate Cord Kit for MET ONE 7000, 24 VDC, 3.7 m (12 ft)
Catalog No. NC3662478 Shop All Beckman Coulter Products
Encompass
Power cord kit for use with Beckman Coulter MET ONE 7000 series remote air particle counters. Provides a 24 VDC connection and a 3.7 m (12 ft) cord length for wall-plate or umbilical routing.
  • 24 VDC power connection.
  • 3.7 m (12 ft) cord length.
  • Provides wall-plate and umbilical routing options.
  • Supplies connectors for DC power termination.
  • Designed for remote air particle counters.
Catalog No. NC3662478 Supplier Beckman Coulter Supplier No. 208876712DC
Power cord kit for use with Beckman Coulter MET ONE 7000 series remote air particle counters. Provides a 24 VDC connection and a 3.7 m (12 ft) cord length for wall-plate or umbilical routing.
  • 24 VDC power connection.
  • 3.7 m (12 ft) cord length.
  • Provides wall-plate and umbilical routing options.
  • Supplies connectors for DC power termination.
  • Designed for remote air particle counters.