Learn More
Grainger Corded Track Saw 55 in Rail 5200 RPM
Catalog No. 502749006
Encompass_Preferred
The Makita Track Saw has a 6-1/2 inch blade diameter and can cut up to 2-3/16 inches at 90 degrees and 1-9/16 inches at 45 degrees. It has a no-load speed of 6,000 RPM and weighs only 14.3 lbs. The saw features an anti-kickback device, electric blade brake, and splinter guard to help control the cut. It is designed for straight, bevel and miter cuts in wood, laminate, veneers and more. The saw can be used with Makita guide rails for longer straight cuts.
Catalog No. 50-274-9006 Supplier Grainger Supplier No. 14F185
The Makita Track Saw has a 6-1/2 inch blade diameter and can cut up to 2-3/16 inches at 90 degrees and 1-9/16 inches at 45 degrees. It has a no-load speed of 6,000 RPM and weighs only 14.3 lbs. The saw features an anti-kickback device, electric blade brake, and splinter guard to help control the cut. It is designed for straight, bevel and miter cuts in wood, laminate, veneers and more. The saw can be used with Makita guide rails for longer straight cuts.