Learn More
Med Vet International Portable Cordless Cautery Pen with High Temp of 2250F
Catalog No. NC9074124
Encompass_Preferred
Small portable cautery, excellent for hemostasis. Uses AA batteries (included). 5/8" fine tip. Cordless and lightweight. High Temp: 2250°F. Intended for coagulating tissue or arresting bleeding from small vessels by heat conducted through the wire tip. Indications include ophthalmic, general and plastic surgery. A protective cap covers the switch and tip. The tip heats in approximately 1.0 second. When, heated to temp, the tip glows a bright red.
Catalog No. NC9074124 Supplier Med Vet International Supplier No. JOR313A
Small portable cautery, excellent for hemostasis. Uses AA batteries (included). 5/8" fine tip. Cordless and lightweight. High Temp: 2250°F. Intended for coagulating tissue or arresting bleeding from small vessels by heat conducted through the wire tip. Indications include ophthalmic, general and plastic surgery. A protective cap covers the switch and tip. The tip heats in approximately 1.0 second. When, heated to temp, the tip glows a bright red.