Learn More
Grainger DEWALT Cordless Dieless Cable Crimping Tool, 750 MCM, Kit
Catalog No. 502759191
Encompass_Preferred
This 20V MAX* cordless dieless crimping tool is designed for crimping #8 to 750 kcmil copper and aluminum cable and connectors. Featuring a universal 4 pusher design, it eliminates the need for manual die changes, while an onboard Crimp Connect® intelligence system tracks crimp cycles, force, calibration, and service history. An integrated LED interface provides real-time status updates on battery, cycle success, and service needs.
  • 360° pivoting head maneuvers in tight spaces
  • Guard rails protect the crimping head when working with small diameter cables
  • Integrated LED light illuminates work areas in low-light conditions
  • Forward and reverse triggers allow for convenient one-handed operation
  • Includes shoulder strap and attachment points for improved jobsite portability
Catalog No. 50-275-9191 Supplier Grainger Supplier No. 165GT6
Add to Cart
Add to Cart
Request a Quote
This 20V MAX* cordless dieless crimping tool is designed for crimping #8 to 750 kcmil copper and aluminum cable and connectors. Featuring a universal 4 pusher design, it eliminates the need for manual die changes, while an onboard Crimp Connect® intelligence system tracks crimp cycles, force, calibration, and service history. An integrated LED interface provides real-time status updates on battery, cycle success, and service needs.
  • 360° pivoting head maneuvers in tight spaces
  • Guard rails protect the crimping head when working with small diameter cables
  • Integrated LED light illuminates work areas in low-light conditions
  • Forward and reverse triggers allow for convenient one-handed operation
  • Includes shoulder strap and attachment points for improved jobsite portability