Learn More
Grainger DeWalt Cordless Cable Crimping Tool, 20V Max, 12-Ton, 750 MCM, 2-Piece Kit
Catalog No. 502759109
Encompass_Preferred
This 20V MAX™ cordless crimping tool delivers 12 tons of force to create UL classified connections on copper and aluminum cables ranging from 8 AWG to 750 kcmil. Featuring a 270° pivoting C-head, this tool is engineered for precision and maneuverability in electrical installations. It includes onboard intelligence that tracks crimping cycles and service requirements, while the Crimp Connect software generates detailed performance reports. Designed for ease of use, the tool incorporates a bright LED work light and accessible forward and reverse triggers for one-handed operation.
  • 12-ton crimping force for versatility
  • Compatible with all industry-standard U-type dies
  • Integrated LED interface for system diagnostics
  • Crimp Connect technology for data reporting
  • 270° pivoting head for enhanced access
  • Bright LED work light
  • Included shoulder strap and kit box
Catalog No. 50-275-9109 Supplier Grainger Supplier No. 165GT5
Add to Cart
Add to Cart
Request a Quote
This 20V MAX™ cordless crimping tool delivers 12 tons of force to create UL classified connections on copper and aluminum cables ranging from 8 AWG to 750 kcmil. Featuring a 270° pivoting C-head, this tool is engineered for precision and maneuverability in electrical installations. It includes onboard intelligence that tracks crimping cycles and service requirements, while the Crimp Connect software generates detailed performance reports. Designed for ease of use, the tool incorporates a bright LED work light and accessible forward and reverse triggers for one-handed operation.
  • 12-ton crimping force for versatility
  • Compatible with all industry-standard U-type dies
  • Integrated LED interface for system diagnostics
  • Crimp Connect technology for data reporting
  • 270° pivoting head for enhanced access
  • Bright LED work light
  • Included shoulder strap and kit box