Learn More
MSC Makita Earth Auger Kit, 40V max XGT, 1 kit
Catalog No. 502819657
Encompass
This cordless earth auger kit provides a powerful, low-maintenance solution for digging even and consistent holes for landscaping, construction, and soil sampling, with zero emissions and reduced noise.
  • Brushless motor delivers power equivalent to a 36cc gas earth auger
  • Automatic Speed Change™ technology adjusts drill speed and torque during drilling
  • Extreme Protection Technology (XPT™) provides increased dust and water resistance
  • Drills holes up to 8 in. wide and 41 in. deep
  • Two speed options (0–400 RPM, 0–1,500 RPM) for power and run time management
  • Reverse switch for clearing auger bit from high-resistance soil
  • Torque limiter reduces gear damage if the bit binds
  • Metal-gear housing increases tool durability
  • Built-in LED light illuminates the work area
Catalog No. 50-281-9657 Supplier MSC Supplier No. 20633921
This cordless earth auger kit provides a powerful, low-maintenance solution for digging even and consistent holes for landscaping, construction, and soil sampling, with zero emissions and reduced noise.
  • Brushless motor delivers power equivalent to a 36cc gas earth auger
  • Automatic Speed Change™ technology adjusts drill speed and torque during drilling
  • Extreme Protection Technology (XPT™) provides increased dust and water resistance
  • Drills holes up to 8 in. wide and 41 in. deep
  • Two speed options (0–400 RPM, 0–1,500 RPM) for power and run time management
  • Reverse switch for clearing auger bit from high-resistance soil
  • Torque limiter reduces gear damage if the bit binds
  • Metal-gear housing increases tool durability
  • Built-in LED light illuminates the work area