Learn More
Research Products International Corp Cordless Engraving Tool
SDP
Catalog No. 501978420
Encompass
Engrave test tubes, bottles, racks, keys, equipment, tools, electronics and more. Simply hold the Engraving Tool like a pen, press the power button, apply light pressure and 'write' to permanently mark most materials including glass, plastic, wood, metal, bone and stone. Marks will not fade if exposed to chemicals or sunlight. Cordless battery power makes it portable for use virtually anywhere indoors or out in the field. Engraving Tool is lightweight and 7 inches long making it easy to store when not in use. Engraving Tool is supplied with an extra engraving tip.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-8420 Supplier Research Products International Corp Supplier No. 247880
Engrave test tubes, bottles, racks, keys, equipment, tools, electronics and more. Simply hold the Engraving Tool like a pen, press the power button, apply light pressure and 'write' to permanently mark most materials including glass, plastic, wood, metal, bone and stone. Marks will not fade if exposed to chemicals or sunlight. Cordless battery power makes it portable for use virtually anywhere indoors or out in the field. Engraving Tool is lightweight and 7 inches long making it easy to store when not in use. Engraving Tool is supplied with an extra engraving tip.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.