Learn More
Stoelting™ Cordless Micro Drill
SDP
Catalog No. 10001013 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Cordless
Catalog No. Model
10-001-013 Cordless
1 options
Catalog No. 10-001-013 Supplier Stoelting™ Supplier No. 58610
Add to Cart
Add to Cart
Stoelting™ Cordless Micro Drill features carbide burr drill bits with a shaft diameter of 2.3 mm and are 4.4 cm in length
Includes:
  • Hard plastic case with die-cut foam for convenient storage
  • Recharger
  • 5 carbide steel burrs with ball sizes of 0.6 mm, 0.8 mm, 1.0 mm, 1.2 mm, and 1.6 mm.
No special tools are necessary to change burrs. Set of 5 assorted burrs available separately.
Specifications
For Use With (Application) Neuroscience
Model Cordless