Learn More
Leica Biosystems Leica Biosystems Cordless Saw Blade, 64 mm - Replacement for Clean-Cut Autopsy Saw and Extractor
Supplier:  Leica Biosystems 380413950
Encompass
Replacement 64 mm cordless saw blade designed as an accessory for cordless autopsy saw and extractor systems. Intended for autopsy and specimen dissection, the compact blade supports precise cutting in confined areas and is compatible with enclosed handpieces used on cordless autopsy saw systems.
  • 64 mm cutting diameter for compact, precise cuts.
  • Intended for autopsy and specimen dissection applications.
  • Compatible with cordless autopsy saw and extractor systems featuring enclosed handpieces.
  • Compact blade profile supports cutting in confined or delicate areas.
  • Provides a direct replacement for worn or damaged blades.
Catalog No. NC3509456