Learn More
MSC Cordova Earplugs, 27 dB NRR, Flanged, 100 Pairs/Pk.
Catalog No. 5000796575
Encompass
These earplugs provide reliable hearing protection in various work environments. Designed for comfort and durability, they feature a metal-detectable cord, making them suitable for industries requiring foreign object detection.
  • Reusable
  • Corded
  • Flanged design
  • Push-in insertion method
  • Metal-detectable
  • Universal size
  • PVC cord material
  • Meets S3.19-1974 standards
Catalog No. 50-007-96575 Supplier MSC Supplier No. 18780718
These earplugs provide reliable hearing protection in various work environments. Designed for comfort and durability, they feature a metal-detectable cord, making them suitable for industries requiring foreign object detection.
  • Reusable
  • Corded
  • Flanged design
  • Push-in insertion method
  • Metal-detectable
  • Universal size
  • PVC cord material
  • Meets S3.19-1974 standards