Learn More
MSC Cordova Gloves, Cut A1, P3, A3, Medium, 12 Pairs
Catalog No. 5000676303
Encompass
These Cor-Touch 6891 gloves offer cut, puncture, and abrasion resistance with ANSI/ISEA ratings of A1, 3, and 3 respectively. Constructed from 13 gauge nylon with a foam nitrile-coated palm, these medium gloves are 9-3/4 in. long and provide a smooth grip.
  • Knit style
  • Foam nitrile palm coating
  • ANSI/ISEA cut resistance level A1
  • ANSI/ISEA puncture resistance level 3
  • ANSI/ISEA abrasion resistance level 3
  • Nylon shell, 13 gauge
  • Overall length 9-3/4 in.
  • Medium size
  • FDA approved
  • Seamless nylon lining
  • Light heat protection
Catalog No. 50-006-76303 Supplier MSC Supplier No. 32769614
These Cor-Touch 6891 gloves offer cut, puncture, and abrasion resistance with ANSI/ISEA ratings of A1, 3, and 3 respectively. Constructed from 13 gauge nylon with a foam nitrile-coated palm, these medium gloves are 9-3/4 in. long and provide a smooth grip.
  • Knit style
  • Foam nitrile palm coating
  • ANSI/ISEA cut resistance level A1
  • ANSI/ISEA puncture resistance level 3
  • ANSI/ISEA abrasion resistance level 3
  • Nylon shell, 13 gauge
  • Overall length 9-3/4 in.
  • Medium size
  • FDA approved
  • Seamless nylon lining
  • Light heat protection