Learn More
MSC Cordova Gloves, Cut A2, Puncture 3, Abrasion 3, 12 Pairs
Catalog No. 5000684143
Encompass
These cut, puncture, and abrasion-resistant gloves feature a 13-gauge HPPE shell with a gray polyurethane palm coating, providing a smooth grip for various industrial applications.
  • Knit style with seamless HPPE lining
  • Polyurethane palm coating for smooth grip
  • ANSI/ISEA Cut Resistance Level A2
  • ANSI/ISEA Puncture Resistance Level 3
  • ANSI/ISEA Abrasion Resistance Level 3
  • Overall length 9 in, cuff length 2-1/2 in
  • CE, EN388, ANSI 105:2016 standards
  • Unisex, large size (numeric 9)
Catalog No. 50-006-84143 Supplier MSC Supplier No. 39040811
These cut, puncture, and abrasion-resistant gloves feature a 13-gauge HPPE shell with a gray polyurethane palm coating, providing a smooth grip for various industrial applications.
  • Knit style with seamless HPPE lining
  • Polyurethane palm coating for smooth grip
  • ANSI/ISEA Cut Resistance Level A2
  • ANSI/ISEA Puncture Resistance Level 3
  • ANSI/ISEA Abrasion Resistance Level 3
  • Overall length 9 in, cuff length 2-1/2 in
  • CE, EN388, ANSI 105:2016 standards
  • Unisex, large size (numeric 9)