Learn More
MSC Cordova Safety Coverall, Microporous, Serged Seams, 3X-Large, 1 Each
Catalog No. 5000792611
Encompass
The DEFENDER microporous, disposable coverall from Cordova Safety is designed to provide an effective barrier against particulates, aerosols, and liquids while maintaining comfort. It is suitable for work environments where hazardous and non-hazardous liquid and dry particulate contaminants may be present.
  • Constructed from tough, composite fabric combining breathable microporous film and soft non-woven polyolefin
  • Features serged seams
  • Zipper front and collar
  • Elastic wrists and ankles
  • ANSI/ISEA 101 compliant sizing
  • Passes ASTM 1670 blood penetration test
  • Anti-static properties
  • Safe for food contact applications
Catalog No. 50-007-92611 Supplier MSC Supplier No. 60113313
The DEFENDER microporous, disposable coverall from Cordova Safety is designed to provide an effective barrier against particulates, aerosols, and liquids while maintaining comfort. It is suitable for work environments where hazardous and non-hazardous liquid and dry particulate contaminants may be present.
  • Constructed from tough, composite fabric combining breathable microporous film and soft non-woven polyolefin
  • Features serged seams
  • Zipper front and collar
  • Elastic wrists and ankles
  • ANSI/ISEA 101 compliant sizing
  • Passes ASTM 1670 blood penetration test
  • Anti-static properties
  • Safe for food contact applications