Learn More
CORDX LABS INC CorDx Tyfast Flu A/B & COVID-19 Multiplex Rapid Test (25PK/CS)
Catalog No. NC3363553
Encompass_Preferred
The CorDx Tyfast Flu A/B & COVID-19 Multiplex Rapid Test is a lateral flow immunoassay intended for the qualitative and differential detection of SARS-CoV-2 and Influenza A and B antigen in an anterior nares sample (from both nostrils). The test provides an efficient and accurate diagnostic solution, giving communities and families fast way to prioritize their health.
Flu A: 87804QW
Flu B: 87804QW-59
Features and Benefits
The First 10-minute test in the US for Flu A/B & COVID-19.
No instrument needed.
High sensitivity, specificity and accuracy
.Production Lead time: Upon volume but in general 1-2 weeks.
Storage: 36-86°F/2~30°C
. Shelf Life: 15 months
CorDx Tyfast Emergency Use Authorization Number(EUA): EUA230055
This product has been authorized only for the detection from SARS-CoV-2, influenza A and influenza B, not for any other viruses or pathogens.
Catalog No. NC3363553 Supplier CORDX LABS INC Supplier No. ACT2100125
The CorDx Tyfast Flu A/B & COVID-19 Multiplex Rapid Test is a lateral flow immunoassay intended for the qualitative and differential detection of SARS-CoV-2 and Influenza A and B antigen in an anterior nares sample (from both nostrils). The test provides an efficient and accurate diagnostic solution, giving communities and families fast way to prioritize their health.
Flu A: 87804QW
Flu B: 87804QW-59
Features and Benefits
The First 10-minute test in the US for Flu A/B & COVID-19.
No instrument needed.
High sensitivity, specificity and accuracy
.Production Lead time: Upon volume but in general 1-2 weeks.
Storage: 36-86°F/2~30°C
. Shelf Life: 15 months
CorDx Tyfast Emergency Use Authorization Number(EUA): EUA230055
This product has been authorized only for the detection from SARS-CoV-2, influenza A and influenza B, not for any other viruses or pathogens.