Learn More
Apexbio Technology LLC Cordycepin 73-03-0 20mg
SDP
Catalog No. 502240084
Encompass_Preferred
Cordycepin (73-03-0) is a small-molecule inhibitor targeting pro-inflammatory cytokines such as IL-1 IL-6 and TNF- It is designed to inhibit inflammatory signaling pathways thereby regulating immune and inflammatory responses Cordycepin exerts its biological activity primarily through interference with RNA synthesis by incorporation into RNA chains leading to premature termination and inhibition of protein translation In cell-based studies cordycepin demonstrates inhibitory activity with IC50 values in the low micromolar range (approximately 5-15 M) depending on experimental conditions and cell models Based on these pharmacological properties cordycepin holds research potential in inflammation regulation immune-response modulation tuberculosis infection modeling and RNA transcription studies
This item is not returnable. View return policy
Catalog No. 50-224-0084 Supplier Apexbio Technology LLC Supplier No. N280820
Cordycepin (73-03-0) is a small-molecule inhibitor targeting pro-inflammatory cytokines such as IL-1 IL-6 and TNF- It is designed to inhibit inflammatory signaling pathways thereby regulating immune and inflammatory responses Cordycepin exerts its biological activity primarily through interference with RNA synthesis by incorporation into RNA chains leading to premature termination and inhibition of protein translation In cell-based studies cordycepin demonstrates inhibitory activity with IC50 values in the low micromolar range (approximately 5-15 M) depending on experimental conditions and cell models Based on these pharmacological properties cordycepin holds research potential in inflammation regulation immune-response modulation tuberculosis infection modeling and RNA transcription studies