Learn More
Selleck Chemical LLC Cordycepin 25mg
SDP
Supplier:  Selleck Chemical LLC S361025MG
Encompass_Preferred
Cordycepin (3'-Deoxyadenosine) is an adenosine analogue, which is readily phosphorylated to its mono-, di-, and triphosphate intracellularly. It has a very potent anti-cancer, anti-oxidant and anti-inflammatory activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3123