Learn More
Cayman Chemical Cordycepin, Purity: ≥98%, CAS: 73-03-0, 100 mg
SDP
Catalog No. 502972645
Encompass_Preferred
A nucleoside analog with diverse biological activities; is converted to cordycepin triphosphate, which inhibits ATP-dependent DNA and RNA synthesis; incorporates into DNA and RNA in SK-HEP-1 cells; is active against B. subtilis at 0.1-1 mg/ml; increases IL-10 secretion and mRNA expression in isolated human PBMCs at 24 µg/ml; increases survival of mice infected with C. albicans or C. krusei at 1 mg/kg per day
Catalog No. 50-297-2645 Supplier Cayman Chemical Supplier No. 14426100 MG
A nucleoside analog with diverse biological activities; is converted to cordycepin triphosphate, which inhibits ATP-dependent DNA and RNA synthesis; incorporates into DNA and RNA in SK-HEP-1 cells; is active against B. subtilis at 0.1-1 mg/ml; increases IL-10 secretion and mRNA expression in isolated human PBMCs at 24 µg/ml; increases survival of mice infected with C. albicans or C. krusei at 1 mg/kg per day