Learn More
Braintree Scientific Core Compact Portable Balance Scale, 600g capacity
SDP
Catalog No. 501954582
Encompass
Core™ Compact Portable Balances Full range tare 9 weighing units Zero tracking Simple 4 button operation Backlit LCD display Stainless steel pan ShockProtect™ overload three-point protection Can be stacked for storage (120mm pan size only) Lock down mounting slot for Kensington™ type lock and cable Dual tare keys Lightweight and portable Modern low profile design Color coded, sealed keypad Low cost and simple to use External calibration Battery power (6 x AA Batteries) Auto sleep/power down function to save battery life AC Adapter **Sold for research purposes only and not for human use**
Catalog No. 50-195-4582 Supplier Braintree Scientific Supplier No. CQT601
Core™ Compact Portable Balances Full range tare 9 weighing units Zero tracking Simple 4 button operation Backlit LCD display Stainless steel pan ShockProtect™ overload three-point protection Can be stacked for storage (120mm pan size only) Lock down mounting slot for Kensington™ type lock and cable Dual tare keys Lightweight and portable Modern low profile design Color coded, sealed keypad Low cost and simple to use External calibration Battery power (6 x AA Batteries) Auto sleep/power down function to save battery life AC Adapter **Sold for research purposes only and not for human use**