Learn More
TrippNT CORE DX CART BLUE SLIDE DOOR
SDP
Supplier:  TrippNT 53375
Encompass_Preferred
TrippNT's Core DX Cart with Global Blue Drawers and Sliding Door organizes, stores, and secures supplies. It offers 3 total locking drawers, 4 shelves, 2 dispensing shelves, and bulk storage with a sliding clear door. Its built-in handles are ergonomic and easy to grab and the sliding door offers a smaller usage footprint. It is constructed from a corrosion-resistant non-metal rust proof polymer with robust 4-inch twin wheel casters capable of easily navigating elevator gaps and uneven surfaces. Available in 6 color variants. Ships fully assembled.
Catalog No. NC3370558