Learn More
TrippNT CORE DX WHT HNG DOOR RAILTOP
SDP
Supplier:  TrippNT 53547
Encompass_Preferred
TrippNT's Core DX Cart with White Drawers, Hinged Door, and Railtop organizes, stores, and secures supplies. It offers a 1-inch railtop for secure top storage, 3 drawers, 4 shelves, 2 dispensing shelves, and bulk storage with a hinged clear door. Its built-in handles are ergonomic and easy to grab. It is constructed from a corrosion-resistant non-metal rust proof polymer with robust 4-inch twin wheel casters capable of easily navigating elevator gaps and uneven surfaces. Available in 6 color variants. Ships fully assembled.
Catalog No. NC3711047