Learn More
SMARTLOGICS LLC Core Gloves, Nitrile - XS - 9 inch Cuff, 3-4 Mil Medical,Chemo-Rated, FDA, Exam Grade, Viral Resistant,Fentanyl tested, Latex-Free & Powder-Free, Ambidextrous, Allergy Free, Blue, Case of 1000
Catalog No. NC3803825
Encompass_Preferred
Nitrile Gloves, Core Gloves, FDA approved, Medical, Examination Grade, Regular cuff - 9 inch cuff - XS3-4 Mil Thickness: Fingertip-Textured for a reliable, non-slip grip on medical instruments; Ambidextrous design.Maximum Barrier Protection - Chemo-Rated: Rigorously tested for use with chemotherapy drugs, providing frontline staff with the highest level of confidence and safety when handling hazardous materials. Exceeds Medical Standards: These gloves are intended for medical purposes and provide an effective barrier to prevent contamination between patient and examiner, making them a trusted choice.Accelerator-Free and 100% Latex-Free : Protects both patients and clinicians from Type I (latex) and Type IV (chemical) allergies. Color: Blue; Critical for use in oncology, emergency medical services, patient examinations, dental offices, where exposure to hazardous substances is a concern, 10 boxes per case (1000 gloves total).
Catalog No. NC3803825 Supplier SMARTLOGICS LLC Supplier No. CGMXS01
Nitrile Gloves, Core Gloves, FDA approved, Medical, Examination Grade, Regular cuff - 9 inch cuff - XS3-4 Mil Thickness: Fingertip-Textured for a reliable, non-slip grip on medical instruments; Ambidextrous design.Maximum Barrier Protection - Chemo-Rated: Rigorously tested for use with chemotherapy drugs, providing frontline staff with the highest level of confidence and safety when handling hazardous materials. Exceeds Medical Standards: These gloves are intended for medical purposes and provide an effective barrier to prevent contamination between patient and examiner, making them a trusted choice.Accelerator-Free and 100% Latex-Free : Protects both patients and clinicians from Type I (latex) and Type IV (chemical) allergies. Color: Blue; Critical for use in oncology, emergency medical services, patient examinations, dental offices, where exposure to hazardous substances is a concern, 10 boxes per case (1000 gloves total).