Learn More
SMARTLOGICS LLC Core Gloves CPE - Small - Disposable Gloves, Food Safe Polyethylene for Food Service & Salon Use, Latex & Allergy Free, Lightweight Embossed, Clear Pack of 2000
Catalog No. NC3803844
Encompass_Preferred
Core Gloves, SMALL - Disposable, Polyethylene (CPE) , Food Service & Multipurpose, Economical Hygiene Protection: An affordable, single-use solution designed for basic hygiene, food handling, and general tasks where frequent glove changes are required , Food Safe Compliant: Conforms to FDA 21 CFR 177.1520 and EU No.10/2011, making them safe for direct contact with food , Allergy-Free Formulation: 100% free from latex, powders, plasticizers, and chemicals, eliminating risks of Type I and Type IV skin allergies , Lightweight 0.92 mil polyethylene resin construction; Non-skin irritant and non-toxic for safe use; Embossed texture for easier handling; Color: Clear; Size: Large , Ideal for food preparation, beauty and salon services, cleaning, housekeeping, painting, gardening, and other general purpose uses , 200 gloves per dispenser box, 30 dispensers per case (2000 gloves total)
Catalog No. NC3803844 Supplier SMARTLOGICS LLC Supplier No. CGNMCLRS00000000001
Core Gloves, SMALL - Disposable, Polyethylene (CPE) , Food Service & Multipurpose, Economical Hygiene Protection: An affordable, single-use solution designed for basic hygiene, food handling, and general tasks where frequent glove changes are required , Food Safe Compliant: Conforms to FDA 21 CFR 177.1520 and EU No.10/2011, making them safe for direct contact with food , Allergy-Free Formulation: 100% free from latex, powders, plasticizers, and chemicals, eliminating risks of Type I and Type IV skin allergies , Lightweight 0.92 mil polyethylene resin construction; Non-skin irritant and non-toxic for safe use; Embossed texture for easier handling; Color: Clear; Size: Large , Ideal for food preparation, beauty and salon services, cleaning, housekeeping, painting, gardening, and other general purpose uses , 200 gloves per dispenser box, 30 dispensers per case (2000 gloves total)