Learn More
Cayman Chemical Core HIstonshumn 100ug
SDP
Catalog No. 502954174
Encompass_Preferred
Source: Highly purified mixture of human core histones (H2A, H2B, H3, and H4) isolated via hydroxyapatite chromatography from HeLa S3 (human cervical adenocarcinoma) nuclear pellet
Catalog No. 50-295-4174 Supplier Cayman Chemical Supplier No. 11010100 UG
Source: Highly purified mixture of human core histones (H2A, H2B, H3, and H4) isolated via hydroxyapatite chromatography from HeLa S3 (human cervical adenocarcinoma) nuclear pellet