Learn More
Adam Equipment Adam Equipment Core Portable Compact Balance CQT2601 2600g capacity x 0.1g readability
Catalog No. NC2612360
Encompass_Preferred
Adam Equipment Core Portable Compact Balances offers outstanding value, providing easy operation and a portable solution for any basic precision weighing tasks. An innovative stackable design saves space and can easily be stored in school supply cupboards
Catalog No. NC2612360 Supplier Adam Equipment Supplier No. CQT 2601
Adam Equipment Core Portable Compact Balances offers outstanding value, providing easy operation and a portable solution for any basic precision weighing tasks. An innovative stackable design saves space and can easily be stored in school supply cupboards