Learn More
Grainger Core Remover Tool Recessed
Catalog No. 502752098
Encompass_Preferred
The Mastercool Valve Core Remover/Installer is a tool for removing and installing valve cores in automotive and HVAC systems. It features a reversible design with a knurled grip handle and a slotted tip to fit standard valve cores. The tool allows quick removal and installation of valve cores without damage for accessing the system to add refrigerant, evacuate the system, or perform repairs. It is made from chrome vanadium steel for durability. This valve core tool covers most common valve core sizes and applications.
Catalog No. 50-275-2098 Supplier Grainger Supplier No. 141C74
The Mastercool Valve Core Remover/Installer is a tool for removing and installing valve cores in automotive and HVAC systems. It features a reversible design with a knurled grip handle and a slotted tip to fit standard valve cores. The tool allows quick removal and installation of valve cores without damage for accessing the system to add refrigerant, evacuate the system, or perform repairs. It is made from chrome vanadium steel for durability. This valve core tool covers most common valve core sizes and applications.