Learn More
BURKLE INC Core Sampler, V4A, length 31.50 in. (800mm), Ø 0.98 in. (25 mm)
Supplier:  BURKLE INC 53100003
Encompass_Preferred
The core drill is the patented solution for the challenging sampling of bulk materials that tend to jam or stick due to their particle shape.
Compared to conventional multi-layer samplers for bulk materials, the two-part core drill has a gap-free design that prevents the particles from tilting during sampling. This guarantees professional and representative sampling through several layers, even with difficult powders and granulates such as glass beads or metal powder.
The areas of application range from sampling from big bags to small containers, with the core sampler demonstrating its strengths in both laboratory and industrial applications.
For particularly dense materials, sampling can be supported by a rubber mallet, which enables the core drill rod to be inserted and removed. The core drill can be easily dismantled for quick and thorough cleaning. The included fastening clip allows the drill rod to be quickly and easily locked into the sampling tube.
Catalog No. 50-000-25650